cis-1,2-Diaminocyclohexane ≥97.0% (by GC, titration analysis)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
C1463.5
C1463.5G
FLUOC1463.5EA
0
GBP
FLUOC1463.5
FLUOC1463.5G
cis-1,2-Diaminocyclohexane ≥97.0% (by GC, titration analysis)
cis-1,2-Diaminocyclohexane
Formula:
C₆H₁₄N₂ Boiling Pt: 93°C Flash Pt: 71°C |
MDL Number:
MFCD00063746 CAS Number: 1436-59-5 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...