N-Boc-ethylenediamine ≥97.0% (by GC)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane
A1371.25G
A1371.14
FLUOA1371.25GEA
0
GBP
FLUOA1371.25G
FLUOA1371.14
N-Boc-ethylenediamine ≥97.0% (by GC)
N-Boc-ethylenediamine
Formula:
C₇H₁₆N₂O₂ MW: 160.22 g/mol Boiling Pt: 105°C |
MDL Number:
MFCD00191871 CAS Number: 57260-73-8 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...