2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Amino-4-hydroxybiphenyl, 3-Amino-[1,1'-biphenyl]-4-ol

A0397 TCA0397.5G
FLUOA0397EA 0 GBP
FLUOA0397 FLUOTCA0397.5G
2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)
2-Amino-4-phenylphenol
Formula: C₁₂H₁₁NO
MW: 185.2200 g/mol
MDL Number: MFCD00059187
CAS Number: 1134-36-7
EINECS: 214-484-8

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR