4-Methyltetrahydropyran 95% stabilised
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Methyltetrahydro-2H-pyran, 4-Methyloxane
Stabilisation:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
467320010
467320025
467321000
ACRO467320010EA
30.3
GBP
ACRO467320010
ACRO467320025
ACRO467321000
4-Methyltetrahydropyran 95% stabilised
4-Methyltetrahydropyran
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 105 °C |
MDL Number:
MFCD07783308 CAS Number: 4717-96-8 EINECS: 225-207-5 UN: 2924 ADR: 3.8 II |
Specification Test Results
Appearance (Color) | Clear colorless |
Color | ≤10 APHA |
Refractive index | 1.4170 to 1.4200 (20°C, 589 nm) |
Appearance (Form) | liquid |
Infrared spectrum | Conforms |
GC | ≥98.0 % |
Stabilizer | 20 ppm BHT |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...