FALGPA
Supplier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIF0744-25MG BIF0744-5MG
APOSBIF0744-25MGEA 141 GBP
APOSBIF0744-25MG APOSBIF0744-5MG
FALGPA
Proteins and Peptides
Substrate for spectrophotometric assay of collagenase