3-Fluoro-4-pyridineboronic acid hydrate
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC9801-1G
PC9801-5G
APOSPC9801-1GEA
104
GBP
APOSPC9801-1G
APOSPC9801-5G
3-Fluoro-4-pyridineboronic acid hydrate
3-Fluoro-4-pyridineboronic acid hydrate
In Suzuki biaryl coupling displacement of boron by an electrophilic species takes place with the formation of a new carbon-carbon or carbon-heteroatom bond.
Formula:
C₅H₅BFNO₂·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD03701686 CAS Number: 1029880-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...