2-Fluoro-6-(tributylstannyl)pyridine 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC8528-1G PC8528-250MG
APOSPC8528-1GEA 314 GBP
APOSPC8528-1G APOSPC8528-250MG
2-Fluoro-6-(tributylstannyl)pyridine 96%
2-Fluoro-6-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
Storage Temperature: Ambient
MDL Number: MFCD09025793
CAS Number: 1025744-38-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR