Fentin fluoride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC7875-1G PC7875-5G
APOSPC7875-1GEA 61 GBP
APOSPC7875-1G APOSPC7875-5G
Fentin fluoride 98%
Fentin fluoride
Formula: C₁₈H₁₅FSn
Melting Pt: >281 °C
Storage Temperature: Ambient
MDL Number: MFCD00015356
CAS Number: 379-52-2
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR