(3,3,3-Trifluoropropyl)dichloromethylsilane 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane

PC7820FB-25G PC7820FB-5G
APOSPC7820FB-25GEA 50.7 GBP
APOSPC7820FB-25G APOSPC7820FB-5G
(3,3,3-Trifluoropropyl)dichloromethylsilane 97%
(3,3,3-Trifluoropropyl)dichloromethylsilane
Formula: C₄H₇Cl₂F₃Si
MW: 211.09 g/mol
Boiling Pt: 121…122 °C
Density: 1.261 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00039294
CAS Number: 675-62-7
EINECS: 211-623-4
UN: 2985
ADR: 3+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR