3,3,3-Trifluoropropan-1-amine hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC7820EF-1G PC7820EF-5G PC7820EF-10G
APOSPC7820EF-1GEA 26.5 GBP
APOSPC7820EF-1G APOSPC7820EF-5G APOSPC7820EF-10G
3,3,3-Trifluoropropan-1-amine hydrochloride 97%
3,3,3-Trifluoropropan-1-amine hydrochloride
Formula: C3H7ClF3N
MW: 149.5426 g/mol
Melting Pt: 222…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00236725
CAS Number: 2968-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR