2',3',5'-Trifluoropropiophenone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC7820B-10G PC7820B-5G PC7820B-1G
APOSPC7820B-10GEA 89.8 GBP
APOSPC7820B-10G APOSPC7820B-5G APOSPC7820B-1G
2',3',5'-Trifluoropropiophenone 98%
2',3',5'-Trifluoropropiophenone
Formula: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD00083545
CAS Number: 243666-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR