3,3,3-Trifluoropropionic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3,3,3-Trifluoropropanoic acid

PC7820A-100G PC7820A-25G PC7820A-5G
APOSPC7820A-100GEA 201 GBP
APOSPC7820A-100G APOSPC7820A-25G APOSPC7820A-5G
3,3,3-Trifluoropropionic acid 98%
3,3,3-Trifluoropropionic acid
Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 146 °C
Melting Pt: 44478 °C
Density: 1.441 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR