3,3,3-Trifluoropropene 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC7820-100G PC7820-500G
APOSPC7820-100GEA 215 GBP
APOSPC7820-100G APOSPC7820-500G
3,3,3-Trifluoropropene 99%
3,3,3-Trifluoropropene
Formula: C₃H₃F₃
Boiling Pt: –21 °C
Density: 0.94 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039265
CAS Number: 677-21-4
EINECS: 211-637-0
UN: 1954
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR