Trifluoromethanesulphonic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Trifluoromethanesulphonic acid

PC7410-1KG PC7410-250G PC7410-500G
APOSPC7410-1KGEA 161 GBP
APOSPC7410-1KG APOSPC7410-250G APOSPC7410-500G
Trifluoromethanesulphonic acid 99%
Trifluoromethanesulphonic acid

Useful catalyst /Extremely strong monoprotic acid.

Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C
Melting Pt: –40 °C
Density: 1.696 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR