1,1,1-Trifluoroethane 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: FC-143a

PC7298-250G PC7298-25G PC7298-100G
APOSPC7298-250GEA 311 GBP
APOSPC7298-250G APOSPC7298-25G APOSPC7298-100G
1,1,1-Trifluoroethane 99%
1,1,1-Trifluoroethane
Formula: C₂H₃F₃
MW: 84.04 g/mol
Boiling Pt: –47 °C
Melting Pt: –111 °C
Density: 0.942 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013575
CAS Number: 420-46-2
UN: 2035
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR