1,2-Difluoroethane
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
FC-152
PC7081-1G
PC7081-5G
APOSPC7081-1GEA
158
GBP
APOSPC7081-1G
APOSPC7081-5G
1,2-Difluoroethane
1,2-Difluoroethane
Formula:
C₂H₄F₂ Boiling Pt: 30.7 °C Density: 0.913 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039286 CAS Number: 624-72-6 UN: 2929 ADR: 6.1+3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...