Heptafluoro-1-methoxypropane 99,5%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Heptafluoropropyl methyl ether

PC7008-250G PC7008-50G
APOSPC7008-250GEA 158 GBP
APOSPC7008-250G APOSPC7008-50G
Heptafluoro-1-methoxypropane 99,5%
Heptafluoro-1-methoxypropane

  • Viscosity 0.471cp@ 23C
  • Surface tension 12.39 dynes/cm.

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 34.18 °C
Melting Pt: –122.5 °C
Density: 1.409 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01320779
CAS Number: 375-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR