1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,1,2,2-Tetrafluoroethyl 2,2,2-trifluoroethyl ether

PC6785-100G PC6785-25G PC6785-5G
APOSPC6785-100GEA 126 GBP
APOSPC6785-100G APOSPC6785-25G APOSPC6785-5G
1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether 99%
1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether
Formula: C₄H₃F₇O
Boiling Pt: 56.7 °C
Melting Pt: –94 °C
Density: 1.4874 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236118
CAS Number: 406-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR