1,1,2,2-Tetrafluoro-1-methoxyethane 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,1,2,2-Tetrafluoroethyl methyl ether

PC6784-100G PC6784-25G
APOSPC6784-100GEA 185 GBP
APOSPC6784-100G APOSPC6784-25G
1,1,2,2-Tetrafluoro-1-methoxyethane 99%
1,1,2,2-Tetrafluoro-1-methoxyethane
Formula: C₃H₄F₄O
MW: 132.06 g/mol
Boiling Pt: 36…37 °C
Melting Pt: –107 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00042106
CAS Number: 425-88-7
EINECS: 207-039-4
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR