1,2,2,2-Tetrafluoroethyl difluoromethyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC6781E-10G PC6781E-25G
APOSPC6781E-10GEA 207 GBP
APOSPC6781E-10G APOSPC6781E-25G
1,2,2,2-Tetrafluoroethyl difluoromethyl ether 97%
1,2,2,2-Tetrafluoroethyl difluoromethyl ether

Useful solvent for dispersing fluorinated materials. Not anaesthetic grade

Formula: C₃H₂F₆O
Boiling Pt: 23…24 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236716
CAS Number: 57041-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR