Perfluoropentane 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Perfluoro-n-pentane, Fluorinert PF 5050, Flutec PP50

PC6564-100G PC6564-25G
APOSPC6564-100GEA 539 GBP
APOSPC6564-100G APOSPC6564-25G
Perfluoropentane 99%
Perfluoropentane
Formula: C₅F₁₂
MW: 288.04 g/mol
Boiling Pt: 29 °C
Melting Pt: –120 °C
Density: 1.664 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042081
CAS Number: 678-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR