Rubidium hexafluoroarsenate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Rubidium hexafluoroarsenate(V)

PC6342-25G PC6342-5G
APOSPC6342-25GEA 406 GBP
APOSPC6342-25G APOSPC6342-5G
Rubidium hexafluoroarsenate 98%
Rubidium hexafluoroarsenate
Formula: AsF₆Rb
MW: 274.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042537
CAS Number: 43023-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR