3-(1,1,2,3,3,3-Hexafluoropropoxy)propene 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Allyl 2H-hexafluoroprop-1-yl ether

PC6210G-25G PC6210G-5G
APOSPC6210G-25GEA 251 GBP
APOSPC6210G-25G APOSPC6210G-5G
3-(1,1,2,3,3,3-Hexafluoropropoxy)propene 97%
3-(1,1,2,3,3,3-Hexafluoropropoxy)propene
Formula: C₆H₆F₆O
MW: 208.1 g/mol
Boiling Pt: 86…89 °C
Density: 1.305 g/cm³ (25 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00043795
CAS Number: 59158-81-5
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR