Perfluoropropane 99,9%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC6210-100G PC6210-25G
APOSPC6210-100GEA 0 GBP
APOSPC6210-100G APOSPC6210-25G
Perfluoropropane 99,9%
Perfluoropropane
Formula: C₃F₈
MW: 188.02 g/mol
Boiling Pt: –39 °C
Melting Pt: –183 °C
Density: 1.352 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039239
CAS Number: 76-19-7
EINECS: 200-941-9
UN: 2424
ADR: 2.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR