1H-Tridecafluorohexane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H-Perfluorohexane

PC6072A-25G PC6072A-5G
APOSPC6072A-25GEA 166 GBP
APOSPC6072A-25G APOSPC6072A-5G
1H-Tridecafluorohexane 98%
1H-Tridecafluorohexane
Formula: C₆HF₁₃
MW: 320.05 g/mol
Boiling Pt: 71 °C
Density: 1.685 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00142632
CAS Number: 355-37-3
EINECS: 206-581-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR