Perfluorohexane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tetradecafluorohexane

PC6072-100G PC6072-25G PC6072-500G
APOSPC6072-100GEA 127 GBP
APOSPC6072-100G APOSPC6072-25G APOSPC6072-500G
Perfluorohexane 98%
Perfluorohexane

Mixed isomers, ca 85% n-isomer. Non-toxic, non-ozone depleting alternative to carbontetrachloride for bromination reaction, ask for reference.

Formula: C₆F₁₄
Boiling Pt: 58…60 °C
Melting Pt: –4 °C
Density: 1.669 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000437
CAS Number: 355-42-0
EINECS: 206-585-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR