1H-Heneicosafluorodecane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H-Perfluorodecane

PC5963-25G PC5963-5G PC5963-1G
APOSPC5963-25GEA 694 GBP
APOSPC5963-25G APOSPC5963-5G APOSPC5963-1G
1H-Heneicosafluorodecane 98%
1H-Heneicosafluorodecane
Formula: C₁₀HF₂₁
Boiling Pt: 159…160 °C
Melting Pt: 31…33 °C
Storage Temperature: Ambient
CAS Number: 375-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR