Perfluorobutane

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC5884-100G PC5884-25G
APOSPC5884-100GEA 660 GBP
APOSPC5884-100G APOSPC5884-25G
Perfluorobutane
Perfluorobutane
Formula: C₄F₁₀
Boiling Pt: –2 °C
Melting Pt: –84.5 °C
Density: 1.517 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042080
CAS Number: 355-25-9
UN: 3163
ADR: 2.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR