Methyl-2,2,3,3,3-pentafluoropropyl ether 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC5789-25G
PC5789-5G
APOSPC5789-25GEA
421
GBP
APOSPC5789-25G
APOSPC5789-5G
Methyl-2,2,3,3,3-pentafluoropropyl ether 97%
Methyl-2,2,3,3,3-pentafluoropropyl ether
Formula:
C₄H₅F₅O Boiling Pt: 46 °C Density: 1.269 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236115 CAS Number: 378-16-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...