2,2,3,3,3-Pentafluoro-1-propanol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC5706-100G PC5706-25G PC5706-500G
APOSPC5706-100GEA 65.5 GBP
APOSPC5706-100G APOSPC5706-25G APOSPC5706-500G
2,2,3,3,3-Pentafluoro-1-propanol 98%
2,2,3,3,3-Pentafluoro-1-propanol

Suitable reagent for the derivatisation of carboxylic acids for ECD and GCMS analysis.

Formula: C₃H₃F₅O
Boiling Pt: 80 °C/748mmHg
Density: 1.505 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004673
CAS Number: 422-05-9
EINECS: 207-012-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR