(2-Fluoro-4-nitrophenyl)methanol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Fluoro-4-nitrobenzyl alcohol

PC57032-1G PC57032-250MG PC57032-5G
APOSPC57032-1GEA 32.8 GBP
APOSPC57032-1G APOSPC57032-250MG APOSPC57032-5G
(2-Fluoro-4-nitrophenyl)methanol 97%
(2-Fluoro-4-nitrophenyl)methanol
Formula: C₇H₆FNO₃
MW: 171.127 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD11110168
CAS Number: 660432-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR