2-Fluoro-4-nitrobenzaldehyde 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC57031-1G PC57031-5G
APOSPC57031-1GEA 35.8 GBP
APOSPC57031-1G APOSPC57031-5G
2-Fluoro-4-nitrobenzaldehyde 97%
2-Fluoro-4-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.10 g/mol
Melting Pt: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00968941
CAS Number: 157701-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR