4-Fluoro-5-methyl-2-nitrophenol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC57030-1G PC57030-500MG PC57030-5G
APOSPC57030-1GEA 266 GBP
APOSPC57030-1G APOSPC57030-500MG APOSPC57030-5G
4-Fluoro-5-methyl-2-nitrophenol 95%
4-Fluoro-5-methyl-2-nitrophenol
Formula: C₇H₆FNO₃
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD19381761
CAS Number: 182880-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR