1,1,2,2,3-Pentafluoropropane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC5703-10G PC5703-25G PC5703-5G
APOSPC5703-10GEA 154 GBP
APOSPC5703-10G APOSPC5703-25G APOSPC5703-5G
1,1,2,2,3-Pentafluoropropane 98%
1,1,2,2,3-Pentafluoropropane
Formula: C₃H₃F₅
Boiling Pt: 25…26 °C
Melting Pt: –82 °C
Density: 1.336 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00153226
CAS Number: 679-86-7
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR