Pentafluoroethane 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC5590-100G
PC5590-400G
APOSPC5590-100GEA
296
GBP
APOSPC5590-100G
APOSPC5590-400G
Pentafluoroethane 99%
Pentafluoroethane
- Solubility in water 0.5%
Formula:
C₂HF₅ Boiling Pt: –49 °C Melting Pt: –103 °C Density: 1.248 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
354-33-6 UN: 3220 ADR: 2.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...