Mercury(II) fluoride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC5045-1G PC5045-5G
APOSPC5045-1GEA 15.9 GBP
APOSPC5045-1G APOSPC5045-5G
Mercury(II) fluoride 97%
Mercury (II) fluoride
Formula: F₂Hg
Melting Pt: 645 °C
Density: 8.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016139
CAS Number: 7783-39-3
UN: 2025
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR