Lithium hexafluoroarsenate 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC4992-50G PC4992-25G
APOSPC4992-50GEA 331 GBP
APOSPC4992-50G APOSPC4992-25G
Lithium hexafluoroarsenate 99%
Lithium hexafluoroarsenate
Formula: AsF₆Li
Storage Temperature: Ambient
MDL Number: MFCD00011076
CAS Number: 29935-35-1
EINECS: 249-963-0
UN: 1557
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR