1,1,2,3,3,3-Hexafluoropropyl methyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC4796-25G PC4796-5G
APOSPC4796-25GEA 73.6 GBP
APOSPC4796-25G APOSPC4796-5G
1,1,2,3,3,3-Hexafluoropropyl methyl ether 97%
1,1,2,3,3,3-Hexafluoropropyl methyl ether
Formula: C₄H₄F₆O
Boiling Pt: 53 °C
Density: 1.419 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041537
CAS Number: 382-34-3
UN: 3271
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR