1,1,1,2,3,3-Hexafluoropropane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H,2H-Perfluoropropane, FC-236ea

PC4787-100G PC4787-25G
APOSPC4787-100GEA 222 GBP
APOSPC4787-100G APOSPC4787-25G
1,1,1,2,3,3-Hexafluoropropane 98%
1,1,1,2,3,3-Hexafluoropropane
Formula: C₃H₂F₆
Boiling Pt: 6 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 431-63-0
EINECS: 207-076-6
UN: 3163
ADR: 2.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR