1,1,1,3,3,3-Hexafluoro-2-propanol 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC4750-1KG PC4750-250G PC4750-25G
APOSPC4750-1KGEA 147 GBP
APOSPC4750-1KG APOSPC4750-250G APOSPC4750-25G
1,1,1,3,3,3-Hexafluoro-2-propanol 99%
1,1,1,3,3,3-Hexafluoro-2-propanol

Versatile intermediate, excellent solvent for polymers, hashigh polarity and low nucleophilicity. Ask for references.

Formula: C₃H₂F₆O
Boiling Pt: 58 °C
Melting Pt: –4 °C
Density: 1.596 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR