3-Fluorophenol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC4080-100G PC4080-25G
APOSPC4080-100GEA 85.6 GBP
APOSPC4080-100G APOSPC4080-25G
3-Fluorophenol 98%
3-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 178 °C
Melting Pt: 44538 °C
Density: 1.238 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR