Trifluoroacetamidine, Technical Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,2,2-Trifluoroacetimidamide, 2,2,2-Trifluoroacetamidine

PC3901-25G PC3901-100G
APOSPC3901-25GEA 105 GBP
APOSPC3901-25G APOSPC3901-100G
Trifluoroacetamidine, Technical Grade
Trifluoroacetamidine

Shelf-life of product is limited

Formula: C₂H₃F₃N₂
MW: 112.05 g/mol
Boiling Pt: 35…37 °C/11mmHg
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00041545
CAS Number: 354-37-0
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR