Ethyl nonafluorobutyl ether (mixture of isomers)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Perfluorobutyl Ethyl Ether, Nonafluorobutyl Ethyl Ether, Ethyl Perfluorobutyl Ether

PC3797-100G PC3797-500G
APOSPC3797-100GEA 36.9 GBP
APOSPC3797-100G APOSPC3797-500G
Ethyl nonafluorobutyl ether (mixture of isomers)
Ethyl nonafluorobutyl ether (mixture of isomers)

Non-ozone depleting solvent, available in bulk

Formula: C₆H₅F₉O
MW: 264.09 g/mol
Boiling Pt: 78 °C
Melting Pt: –138 °C
Density: 1.43 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 813458-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR