2,2-Difluoroethyl trifluoromethyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC3794-1G PC3794-5G
APOSPC3794-1GEA 165 GBP
APOSPC3794-1G APOSPC3794-5G
2,2-Difluoroethyl trifluoromethyl ether 97%
2,2-Difluoroethyl trifluoromethyl ether
Formula: C₃H₃F₅O
Boiling Pt: 26 °C
Density: 1.375 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093310
CAS Number: 84011-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR