Ethyl-1,1,2,2-tetrafluoroethyl ether

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC3265-100G PC3265-25G
APOSPC3265-100GEA 196 GBP
APOSPC3265-100G APOSPC3265-25G
Ethyl-1,1,2,2-tetrafluoroethyl ether
Ethyl-1,1,2,2-tetrafluoroethyl ether
Formula: C₄H₆F₄O
MW: 146.08 g/mol
Boiling Pt: 57 °C
Density: 1.198 g/cm³ (25 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00069162
CAS Number: 512-51-6
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR