Fluoroethane 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ethyl fluoride

PC3206-100G PC3206-10G PC3206-25G
APOSPC3206-100GEA 563 GBP
APOSPC3206-100G APOSPC3206-10G APOSPC3206-25G
Fluoroethane 97%
Fluoroethane
Formula: C₂H₅F
MW: 48.06 g/mol
Boiling Pt: –37 °C
Melting Pt: –143.2 °C
Density: 0.818 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 353-36-6
EINECS: 206-531-6
UN: 2453
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR