Difluoromethyl-2,2,2-trifluoroethyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC2849C-25G PC2849C-100G PC2849C-5G
APOSPC2849C-25GEA 341 GBP
APOSPC2849C-25G APOSPC2849C-100G APOSPC2849C-5G
Difluoromethyl-2,2,2-trifluoroethyl ether 97%
Difluoromethyl-2,2,2-trifluoroethyl ether
Formula: C₃H₃F₅O
Boiling Pt: 28…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00153127
CAS Number: 1885-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR