1,1-Difluoroethane 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC2800-100G
PC2800-250G
APOSPC2800-100GEA
291
GBP
APOSPC2800-100G
APOSPC2800-250G
1,1-Difluoroethane 99%
1,1-Difluoroethane
Formula:
C₂H₄F₂ Boiling Pt: –25 °C Melting Pt: –117 °C Density: 0.966 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000449 CAS Number: 75-37-6 UN: 1030 ADR: 2.1, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...