5-Fluoronicotinic acid 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC2505-25G
PC2505-5G
APOSPC2505-25GEA
52.5
GBP
APOSPC2505-25G
APOSPC2505-5G
5-Fluoronicotinic acid 97%
5-Fluoronicotinic acid
Formula:
C₆H₄FNO₂ MW: 141.10 g/mol Melting Pt: 193…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318537 CAS Number: 402-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...