Isoflurane 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC2045-25G
PC2045-100G
APOSPC2045-25GEA
58.1
GBP
APOSPC2045-25G
APOSPC2045-100G
Isoflurane 99%
Isoflurane
Useful solvent for dispersing fluorinated materials.Pharmacology, Byles et al., Can. Anaesth. Soc. J., 1971, 35, 8-53. *Not anaesthetic grade*
Formula:
C₃H₂ClF₅O Boiling Pt: 48.5 °C Density: 1.496 g/cm³ (25 °C) Flash Pt: >61.11 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066609 CAS Number: 26675-46-7 EINECS: 247-897-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...